missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ CLA IdentiFiler™ Plus PCR Amplification Kit

Catalog No. A65672
Change view
Click to view available options
Quantity:
200 x 25 μL Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A65672 200 x 25 μL Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A65672 Supplier Applied Biosystems™ Supplier No. A65672

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The CLA (Cell Line Authentication) IdentiFiler Plus PCR Amplification Kit simplifies the amplification step during authentication of cell lines.

The CLA (Cell Line Authentication) IdentiFiler Plus PCR Amplification Kit simplifies the amplification step during authentication of cell lines. It includes the same primers and allelic ladder as the widely used AmpFLSTR Identifiler Plus kit and harnesses next-generation PCR amplification technology to help provide enhanced sensitivity, a cleaner baseline, improved performance with mixtures, and the ability to overcome high levels of PCR inhibition. The kit includes GeneScan 600 LIZ Size Standard v2.0, a five-dye-labeled, high-density size standard for the reproducible sizing of fragment analysis data.

Features of the CLA IdentiFiler Plus PCR Amplification Kit include:

  • 5-dye, 16-locus STR kit compatible with all cell line databasing loci standards
  • Improved version of legacy AmpFLSTR Identifiler Plus kit
  • Size standard included for ease of purchase
  • Compatible with SeqStudio, 3730/xL, and 3500/xL genetic analyzers

Cell line authentication using short tandem repeat (STR) analysis kits can be used both for human sample authentication (HSA) and mixed sample analysis (MSA). In HSA, there is a need to verify the DNA profile of the sample to check for sample cross-contamination or sample mix-up, or to perform a quality check. For MSA there is a need to deconvolute the presence of multiple contributors in a sample (chimerism) through identification of multiple DNA genotypes in the sample.

Specifications

Content And Storage • AmpFLSTR Identifiller Plus PCR Ampl Kit
• GS Liz600 Size Standard
Detection Method Gel Electrophoresis
Form Liquid
Format Kit
GC-Rich PCR Performance High
PCR Method End-point PCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed 60 min.
Technique STR (Short Tandem Repeats) Analysis
For Use With (Equipment) SeqStudio Flex 3500, SeqStudio 3730xl Genetic Analyzers
Kit Type Multicomponent
Label or Dye LIZ, FAM, NED, PET, VIC
Quantity 200 x 25 μL Reactions
Species Human
Target Organism Class Human cell lines
For Use With (Application) Genotyping, Cell Analysis
Purification Target Cell DNA
Reaction Format Tube
Sample Type Genomic DNA
Starting Material Cell Line Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.