Learn More
Applied Biosystems™ CLA IdentiFiler™ Direct PCR Amplification Kit
Description
The CLA (Cell Line Authentication) IdentiFiler Direct PCR Amplification Kit simplifies the amplification step during authentication of cell lines. The kit enables direct amplification of the same 16 loci included in our AmpFLSTR Identifiler Direct kit, eliminating the need to perform DNA extraction or purification steps for DNA database and other single-source reference samples. Blood or buccal samples on FTA cards can be punched into PCR plates or tubes, and taken directly to PCR amplification, without reduction in data quality. Samples collected using a Buccal DNA Collector or other non-FTA substrate can be processed without heat incubation or additional workflow steps using Prep-n-Go Buffer (available separately). The CLA Identifiler kit includes GeneScan 600 LIZ Size Standard v2.0, a five-dye-labeled, high-density size standard for the reproducible sizing of fragment analysis data.
Features of the CLA IdentiFiler Direct PCR Amplification Kit include:
- 5-dye, 16-locus STR analysis kit compatible with all cell line databasing loci standardsImproved version of legacy AmpFLSTR Identifiler Direct kit
- Size standard included for ease of purchase
- Compatible with SeqStudio, 3730/xL, and 3500/xL genetic analyzers
Cell line authentication using short tandem repeat (STR) analysis kits can be used both for human sample authentication (HSA) and mixed sample analysis (MSA). In HSA, there is a need to verify the DNA profile of the sample to check for sample cross-contamination or sample mix-up, or to perform a quality check. For MSA there is a need to deconvolute the presence of multiple contributors in a sample (chimerism) through identification of multiple DNA genotypes in the sample.
Specifications
Specifications
| Content And Storage | • IdentiFiler Direct PCR Amplification Kit • Liz600 Size Standard |
| Detection Method | Gel Electrophoresis |
| Form | Liquid |
| Format | Kit |
| GC-Rich PCR Performance | High |
| PCR Method | Direct PCR |
| Polymerase | AmpliTaq Gold™ DNA Polymerase |
| Reaction Speed | 60 min. |
| Technique | STR (Short Tandem Repeats) Analysis |
| For Use With (Equipment) | SeqStudio Flex™ 3500, SeqStudio™ 3730xl Genetic Analyzers |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.