missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, Monohydrate, Granular, FCC, 99.5-100.5%, Spectrum™ Chemical
C1283, 5949-29-1, C6H8O7·H2O
Supplier: Spectrum Chemical Mfg Cor C128312KG
Description
Spectrum™ Chemical Citric Acid, Monohydrate, Granular, FCC is widely used as acidity regulator and presevative in food production and as an acidity regulator and antioxidant in beverages. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 5949-29-1 | |
| C6H10O8 | |
| O.OC(=O)CC(O)(CC(O)=O)C(O)=O | |
| 210.14 | |
| FCC | |
| 0.0005 |
| 1 | |
| YASYEJJMZJALEJ-UHFFFAOYSA-N | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate | |
| 99.5 to 100.5% | |
| 12 kg | |
| Poly Pail |
Chemical Identifiers
| 5949-29-1 | |
| 210.14 | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate |
| C6H10O8 | |
| YASYEJJMZJALEJ-UHFFFAOYSA-N | |
| O.OC(=O)CC(O)(CC(O)=O)C(O)=O |