missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Monohydrate, Granular, FCC, 99.5-100.5%, Spectrum™ Chemical

Catalog No. p-7157146
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601725 500 g Poly Bottle
18601726 2.5 kg Poly Pail
18601727 12 kg Poly Pail
18601728 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601725 Supplier Spectrum Chemical Mfg Cor Supplier No. C1283500GM
Only null left

C1283, 5949-29-1, C6H8O7·H2O

Spectrum™ Chemical Citric Acid, Monohydrate, Granular, FCC is widely used as acidity regulator and presevative in food production and as an acidity regulator and antioxidant in beverages. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

CAS Max % 1
Percent Purity 99.5 to 100.5%
Grade FCC
Quantity 500 g
Ignition Residue 0.0005
Packaging Poly Bottle