Learn More
Thermo Scientific Chemicals cis-5,8,11,14,17-Eicosapentaenoic acid
CAS: 10417-94-4 | C20H30O2, C20H30O2 | 302.46 g/mol
$422.36 - $825.03
Chemical Identifiers
| CAS | 10417-94-4 |
|---|---|
| Molecular Formula | C20H30O2 |
| Synonym | EPA; Timnodonic acid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ66060MC
|
Thermo Scientific Chemicals
J66060MC |
100 mg |
Each for $422.36
|
|
|||||
|
AAJ66060ME
|
Thermo Scientific Chemicals
J66060ME |
500 mg |
Each for $825.03
|
|
|||||
Description
Important polyunsaturated fatty acid of the marine food chain that serves as a precursor for the prostaglandin-3 and thromboxane-3 families. It differs from arachidonic acid (the eicosatetraenoic acid that is a precursor for the prostaglandin and thromboxane-2 families) by the extra double bond between the third and fourth carbons from the methyl end of the molecule. Antilipemic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10417-94-4 | |
| C20H30O2 | |
| EPA; Timnodonic acid |
Specifications
| Colorless | |
| 10417-94-4 | |
| 302.45 | |
| 93°C (199°F) | |
| Air and Light Sensitive | |
| Soluble in methanol | |
| Oil | |
| JAZBEHYOTPTENJ-JLNKQSITSA-N | |
| (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoic acid |
| -30°C to -10°C | |
| C20H30O2 | |
| EPA; Timnodonic acid | |
| 0.943 | |
| -54° | |
| cis-5,8,11,14,17-Eicosapentaenoic acid | |
| 1714433 | |
| CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(O)=O | |
| 302.46 |
Safety and Handling
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only