accessibility menu, dialog, popup

UserName

CAS RN 10417-94-4

IUPAC Name: (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoic acid
Synonyms: eicosapentaenoic acid 二十碳五烯酸 timnodonic acid eicosapentaenoate timnodonate EPA icosapento icosapentum all-cis-icosapentaenoate icosapent
Molecular Weight (g/mol): 302.46
Molecular Formula: C20H30O2
InChi Key: JAZBEHYOTPTENJ-JLNKQSITSA-N
SMILES: CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(O)=O
Percent Purity 
  • (2)
Product Type 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results