missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals cis-5,8,11,14,17-Eicosapentaenoic acid

Catalog No. AAJ66060ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66060ME 500 mg
AAJ66060MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66060ME Supplier Thermo Scientific Chemicals Supplier No. J66060ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10417-94-4 | C20H30O2, C20H30O2 | 302.46 g/mol

Important polyunsaturated fatty acid of the marine food chain that serves as a precursor for the prostaglandin-3 and thromboxane-3 families. It differs from arachidonic acid (the eicosatetraenoic acid that is a precursor for the prostaglandin and thromboxane-2 families) by the extra double bond between the third and fourth carbons from the methyl end of the molecule. Antilipemic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10417-94-4
Molecular Formula C20H30O2
Synonym EPA; Timnodonic acid

Specifications

Quantity 500 mg
Description Colorless
Content And Storage -30°C to -10°C
Formula Weight 302.45
Flash Point 93°C (199°F)
Density 0.943
Sensitivity Air and Light Sensitive
Melting Point -54°
Solubility Soluble in methanol
Product Type cis-5,8,11,14,17-Eicosapentaenoic acid
Form Oil
Show More Show Less

RUO – Research Use Only