Learn More
cis-4-Isopropylcyclohexanecarboxylic acid, 97%
CAS: 7084-93-7 | C10H18O2 | 170.25 g/mol
Supplier: Thermo Scientific Chemicals H6259403
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C10H18O2 | |
| MFCD04004145,MFCD01734696,MFCD19706019 | |
| trans-4-isopropylcyclohexanecarboxylic acid, trans-4-isopropylcyclohexane carboxylic acid, 4-isopropylcyclohexanecarboxylic acid, hexahydrocumic acid, isopropyl-cyclohexanecarboxylic acid, cis-4-isopropylcyclohexanecarboxylic acid, p-isopropylhexahydrobenzoic acid, cyclohexanecarboxylic acid, 4-1-methylethyl, cyclohexanecarboxylic acid, 4-isopropyl, cyclohexanecarboxylic acid, 4-1-methylethyl-, trans | |
| 4-propan-2-ylcyclohexane-1-carboxylic acid |
Specifications
| cis-4-Isopropylcyclohexanecarboxylic acid | |
| C10H18O2 | |
| 1 g | |
| YRQKWRUZZCBSIG-UHFFFAOYSA-N | |
| 4-propan-2-ylcyclohexane-1-carboxylic acid | |
| 81526 | |
| 97% |
| 7084-93-7 | |
| MFCD04004145,MFCD01734696,MFCD19706019 | |
| trans-4-isopropylcyclohexanecarboxylic acid, trans-4-isopropylcyclohexane carboxylic acid, 4-isopropylcyclohexanecarboxylic acid, hexahydrocumic acid, isopropyl-cyclohexanecarboxylic acid, cis-4-isopropylcyclohexanecarboxylic acid, p-isopropylhexahydrobenzoic acid, cyclohexanecarboxylic acid, 4-1-methylethyl, cyclohexanecarboxylic acid, 4-isopropyl, cyclohexanecarboxylic acid, 4-1-methylethyl-, trans | |
| CC(C)C1CCC(CC1)C(O)=O | |
| 170.25 | |
| 170.24 | |
| Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures