missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Isopropylcyclohexanecarboxylic acid, 97%

Catalog No. p-7054408
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62594MD 250 mg
AAH6259403 1 g
AAH6259406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62594MD Supplier Thermo Scientific Chemicals Supplier No. H62594MD

CAS: 7084-93-7 | C10H18O2 | 170.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7084-93-7
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD04004145,MFCD01734696,MFCD19706019
InChI Key YRQKWRUZZCBSIG-UHFFFAOYSA-N
Synonym trans-4-isopropylcyclohexanecarboxylic acid, trans-4-isopropylcyclohexane carboxylic acid, 4-isopropylcyclohexanecarboxylic acid, hexahydrocumic acid, isopropyl-cyclohexanecarboxylic acid, cis-4-isopropylcyclohexanecarboxylic acid, p-isopropylhexahydrobenzoic acid, cyclohexanecarboxylic acid, 4-1-methylethyl, cyclohexanecarboxylic acid, 4-isopropyl, cyclohexanecarboxylic acid, 4-1-methylethyl-, trans
PubChem CID 81526
IUPAC Name 4-propan-2-ylcyclohexane-1-carboxylic acid
SMILES CC(C)C1CCC(CC1)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 170.24
Percent Purity 97%
Physical Form Solid
Chemical Name or Material cis-4-Isopropylcyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only