CAS RN 7084-93-7
CAS RN 7084-93-7
IUPAC Name:
4-(propan-2-yl)cyclohexane-1-carboxylic acid
Synonyms:
4-isopropylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
170.25
Molecular Formula:
C10H18O2
InChi Key:
YRQKWRUZZCBSIG-UHFFFAOYSA-N
SMILES:
CC(C)C1CCC(CC1)C(O)=O
1
–
1
of
1
results
1
cis-4-Isopropylcyclohexanecarboxylic acid, 97%
CAS: 7084-93-7 Molecular Formula: C10H18O2 Molecular Weight (g/mol): 170.25 MDL Number: MFCD04004145,MFCD01734696,MFCD19706019 InChI Key: YRQKWRUZZCBSIG-UHFFFAOYSA-N Synonym: trans-4-isopropylcyclohexanecarboxylic acid,trans-4-isopropylcyclohexane carboxylic acid,4-isopropylcyclohexanecarboxylic acid,hexahydrocumic acid,isopropyl-cyclohexanecarboxylic acid,cis-4-isopropylcyclohexanecarboxylic acid,p-isopropylhexahydrobenzoic acid,cyclohexanecarboxylic acid, 4-1-methylethyl,cyclohexanecarboxylic acid, 4-isopropyl,cyclohexanecarboxylic acid, 4-1-methylethyl-, trans PubChem CID: 81526 IUPAC Name: 4-propan-2-ylcyclohexane-1-carboxylic acid SMILES: CC(C)C1CCC(CC1)C(O)=O