missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-4-Hexen-1-ol, 97%
CAS: 928-91-6 | C6H12O | 100.16 g/mol
$341.23
Chemical Identifiers
| CAS | 928-91-6 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.16 |
| MDL Number | MFCD00148974,MFCD00009713 |
| InChI Key | VTIODUHBZHNXFP-IHWYPQMZSA-N |
| Synonym | cis-4-hexen-1-ol, z-hex-4-en-1-ol, z-4-hexen-1-ol, 4z-hex-4-en-1-ol, 4-hexen-1-ol, z, unii-780wma6j84, e-4-hexenol, cis-1-hydroxy-4-hexene, cis-4-hexene-1-ol, 4z-4-hexen-1-ol # |
| PubChem CID | 5365589 |
| IUPAC Name | (Z)-hex-4-en-1-ol |
| SMILES | C\C=C/CCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1926614
|
N/A | 25 g |
Each for $341.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C6H12O | |
| MFCD00148974,MFCD00009713 | |
| cis-4-hexen-1-ol, z-hex-4-en-1-ol, z-4-hexen-1-ol, 4z-hex-4-en-1-ol, 4-hexen-1-ol, z, unii-780wma6j84, e-4-hexenol, cis-1-hydroxy-4-hexene, cis-4-hexene-1-ol, 4z-4-hexen-1-ol # | |
| (Z)-hex-4-en-1-ol |
Specifications
| 928-91-6 | |
| 158°C | |
| Pleasant | |
| MFCD00148974,MFCD00009713 | |
| cis-4-hexen-1-ol, z-hex-4-en-1-ol, z-4-hexen-1-ol, 4z-hex-4-en-1-ol, 4-hexen-1-ol, z, unii-780wma6j84, e-4-hexenol, cis-1-hydroxy-4-hexene, cis-4-hexene-1-ol, 4z-4-hexen-1-ol # | |
| C\C=C/CCCO | |
| 100.16 | |
| 100.16 | |
| cis-4-Hexen-1-ol |
| 0.857 | |
| 65°C (149°F) | |
| C6H12O | |
| UN1987 | |
| VTIODUHBZHNXFP-IHWYPQMZSA-N | |
| (Z)-hex-4-en-1-ol | |
| 5365589 | |
| 97% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P280-P370+P378q-P501c
H227
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 213-187-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only