accessibility menu, dialog, popup

UserName

CH3OH(Z)

CAS RN 928-91-6

IUPAC Name: (4Z)-hex-4-en-1-ol
Synonyms: (4Z)-hex-4-en-1-ol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: VTIODUHBZHNXFP-IHWYPQMZSA-N
SMILES: C\C=C/CCCO
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)