missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hexen-1-ol, 97%

Catalog No. AAA1926614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1926614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1926614 Supplier Thermo Scientific Chemicals Supplier No. A1926614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 928-91-6 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 928-91-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00148974,MFCD00009713
InChI Key VTIODUHBZHNXFP-IHWYPQMZSA-N
Synonym cis-4-hexen-1-ol, z-hex-4-en-1-ol, z-4-hexen-1-ol, 4z-hex-4-en-1-ol, 4-hexen-1-ol, z, unii-780wma6j84, e-4-hexenol, cis-1-hydroxy-4-hexene, cis-4-hexene-1-ol, 4z-4-hexen-1-ol #
PubChem CID 5365589
IUPAC Name (Z)-hex-4-en-1-ol
SMILES C\C=C/CCCO

Specifications

Density 0.857
Boiling Point 158°C
Flash Point 65°C (149°F)
Odor Pleasant
Quantity 25 g
UN Number UN1987
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material cis-4-Hexen-1-ol
Hazard Category H227
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 213-187-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only