missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%

CAS: 6217-54-5 | C22H32O2 | 328.49 g/mol

$262.49 - $888.16

Chemical Identifiers

CAS 6217-54-5
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.49
InChI Key MBMBGCFOFBJSGT-OBOJEMQYSA-N
PubChem CID 57417355
IUPAC Name (4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
SMILES CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC427641000
View Documents Promotion Details
Thermo Scientific Chemicals
427641000
100 mg Glass Bottle
Each for $262.49
Only null left
 
AC427640010
View Documents Promotion Details
Thermo Scientific Chemicals
427640010
1 g Glass bottle
Each for $888.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6217-54-5
328.49
57417355
CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O
C22H32O2
MBMBGCFOFBJSGT-OBOJEMQYSA-N
(4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid

Specifications

6217-54-5
Colorless to Yellow
62°C
97.5% min. (GC)
CH3(CH2CHCH)6CH2CH2COOH
15, 3443
MBMBGCFOFBJSGT-OBOJEMQYSA-N
(4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
57417355
98%
cis-4,7,10,13,16,19-Docosahexaenoic acid
-44°C
0.9500g/mL
Authentic
C22H32O2
0.95
cis-4,7,10,13,16,19-docosahexaenoic acid, 4,7,10,13,16,19-cis-docosahexaenoic acid, cis-4, 7, 10, 13, 16, 19-docosahexaenoic acid, 4z-docosa-4,7,10,13,16,19-hexaenoic acid
CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O
328.49
328.49
Oil
Videos
SDS
Documents

Documents

RUO – Research Use Only