missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%

CAS: 6217-54-5 | C22H32O2 | 328.49 g/mol

Supplier:  Thermo Scientific Chemicals 427640010

Quantity 1 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC427640010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6217-54-5
328.49
57417355
CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O
C22H32O2
MBMBGCFOFBJSGT-OBOJEMQYSA-N
(4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid

Specifications

cis-4,7,10,13,16,19-Docosahexaenoic acid
-44°C
0.9500g/mL
Authentic
Glass bottle
CH3(CH2CHCH)6CH2CH2COOH
0.95
cis-4,7,10,13,16,19-docosahexaenoic acid, 4,7,10,13,16,19-cis-docosahexaenoic acid, cis-4, 7, 10, 13, 16, 19-docosahexaenoic acid, 4z-docosa-4,7,10,13,16,19-hexaenoic acid
CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O
328.49
328.49
Oil
6217-54-5
Colorless to Yellow
62°C
97.5% min. (GC)
C22H32O2
1 g
15, 3443
MBMBGCFOFBJSGT-OBOJEMQYSA-N
(4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
57417355
98%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions