missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Nonen-1-ol, 95%

CAS: 41453-56-9 | C9H18O | 142.242 g/mol

$112.12 - $395.19

Chemical Identifiers

CAS 41453-56-9
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00063210
InChI Key NSSALFVIQPAIQK-FPLPWBNLSA-N
Synonym cis-2-nonen-1-ol, z-2-nonen-1-ol, z-non-2-en-1-ol, unii-jlw774vc2r, 2-nonen-1-ol, z, 2z-non-2-en-1-ol, 2z-2-nonen-1-ol, jlw774vc2r, fema no. 3720, z-2-nonenol
PubChem CID 5365027
IUPAC Name (Z)-non-2-en-1-ol
SMILES CCCCCCC=CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1884206
View Documents Promotion Details
N/A 5 g
Each for $112.12
Only null left
 
AAA1884214
View Documents Promotion Details
N/A 25 g
Each for $395.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

41453-56-9
142.242
NSSALFVIQPAIQK-FPLPWBNLSA-N
5365027
CCCCCCC=CCO
C9H18O
MFCD00063210
cis-2-nonen-1-ol, z-2-nonen-1-ol, z-non-2-en-1-ol, unii-jlw774vc2r, 2-nonen-1-ol, z, 2z-non-2-en-1-ol, 2z-2-nonen-1-ol, jlw774vc2r, fema no. 3720, z-2-nonenol
(Z)-non-2-en-1-ol

Specifications

41453-56-9
106°C (20 mmHg)
C9H18O
MFCD00063210
NSSALFVIQPAIQK-FPLPWBNLSA-N
(Z)-non-2-en-1-ol
5365027
95%
0.845
101°C (214°F)
1.45
cis-2-nonen-1-ol, z-2-nonen-1-ol, z-non-2-en-1-ol, unii-jlw774vc2r, 2-nonen-1-ol, z, 2z-non-2-en-1-ol, 2z-2-nonen-1-ol, jlw774vc2r, fema no. 3720, z-2-nonenol
CCCCCCC=CCO
142.242
142.24
cis-2-Nonen-1-ol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 255-376-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only