missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Nonen-1-ol, 95%

Catalog No. AAA1884206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1884206 5 g
AAA1884214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1884206 Supplier Thermo Scientific Chemicals Supplier No. A1884206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 41453-56-9 | C9H18O | 142.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41453-56-9
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00063210
InChI Key NSSALFVIQPAIQK-FPLPWBNLSA-N
Synonym cis-2-nonen-1-ol, z-2-nonen-1-ol, z-non-2-en-1-ol, unii-jlw774vc2r, 2-nonen-1-ol, z, 2z-non-2-en-1-ol, 2z-2-nonen-1-ol, jlw774vc2r, fema no. 3720, z-2-nonenol
PubChem CID 5365027
IUPAC Name (Z)-non-2-en-1-ol
SMILES CCCCCCC=CCO

Specifications

Density 0.845
Boiling Point 106°C (20 mmHg)
Flash Point 101°C (214°F)
Refractive Index 1.45
Quantity 5 g
Formula Weight 142.24
Percent Purity 95%
Chemical Name or Material cis-2-Nonen-1-ol
EINECSNumber 255-376-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only