accessibility menu, dialog, popup

UserName

H3COH(Z)

CAS RN 41453-56-9

IUPAC Name: (2Z)-non-2-en-1-ol
Synonyms: (2Z)-non-2-en-1-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: NSSALFVIQPAIQK-FPLPWBNLSA-N
SMILES: CCCCCC\C=C/CO
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)