missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CIQ

Catalog No. 410550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
410550 50 mg
410510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 410550 Supplier Tocris Bioscience™ Supplier No. 4105/50
Only null left

Potentiator of NMDA receptors containing GluN2C/GluN2D

Subunit-selective potentiator of NMDA receptors containing the GluN2C (formally NR2C) or GluN2D (formally NR2D) subunit. Increases channel opening efficiency and enhances NMDA receptor responses.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 486427-17-2
Molecular Formula C26H26ClNO5

Specifications

Quantity 50 mg
Formula Weight 467.94
Purity >98%
Molecular Weight (g/mol) 467.946
Chemical Name or Material CIQ
Recommended Storage Store at +4°C