missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ciprofibrate 98.0+%, TCI America™
$315.03 - $788.49
Chemical Identifiers
| CAS | 52214-84-3 |
|---|---|
| Molecular Formula | C13H14Cl2O3 |
| Molecular Weight (g/mol) | 289.152 |
| MDL Number | MFCD00467135 |
| InChI Key | KPSRODZRAIWAKH-UHFFFAOYSA-N |
| Synonym | 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropionic Acid |
| PubChem CID | 2763 |
| ChEBI | CHEBI:50867 |
| IUPAC Name | 2-[4-(2,2-dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid |
| SMILES | CC(C)(C(=O)O)OC1=CC=C(C=C1)C2CC2(Cl)Cl |
Chemical Identifiers
| 52214-84-3 | |
| 289.152 | |
| KPSRODZRAIWAKH-UHFFFAOYSA-N | |
| 2763 | |
| 2-[4-(2,2-dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid |
| C13H14Cl2O3 | |
| MFCD00467135 | |
| 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropionic Acid | |
| CHEBI:50867 | |
| CC(C)(C(=O)O)OC1=CC=C(C=C1)C2CC2(Cl)Cl |
Specifications
| 52214-84-3 | |
| White-Yellow | |
| MFCD00467135 | |
| 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropionic Acid | |
| CC(C)(C(=O)O)OC1=CC=C(C=C1)C2CC2(Cl)Cl | |
| 289.152 | |
| CHEBI:50867 | |
| ≥98.0% (GC,T) | |
| Ciprofibrate |
| 118°C | |
| C13H14Cl2O3 | |
| 1 g | |
| KPSRODZRAIWAKH-UHFFFAOYSA-N | |
| 2-[4-(2,2-dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid | |
| 2763 | |
| 289.15 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UF0880000
TSCA : No
Recommended Storage : Refrigerator