accessibility menu, dialog, popup

UserName

H3CCH3OClClOHO

CAS RN 52214-84-3

IUPAC Name: 2-[4-(2,2-dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid
Synonyms: ciprofibrate ciprofibratum modalim ciprofibrato 环丙贝特 sanofi winthrop brand of ciprofibrate ciprofibrate (product) lipanor sanofi synthelabo brand of ciprofibrate ccris 173
Molecular Weight (g/mol): 289.15
Molecular Formula: C13H14Cl2O3
InChi Key: KPSRODZRAIWAKH-UHFFFAOYNA-N
SMILES: CC(C)(OC1=CC=C(C=C1)C1CC1(Cl)Cl)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results