missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CINP Monoclonal Antibody (K70038_12G9), Invitrogen™
Mouse Monoclonal Antibody
$1,601.06 - $2,821.92
Specifications
| Antigen | CINP |
|---|---|
| Clone | K70038_12G9 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: MEAKTLGTVT PRKPVLSVSA RKIKDNAADW HNLILKWETL NDAGFTTANN IANLKISLLN KDKIELDSSS PASKENEEKV CLEYNEELEK LCEELQATLD GLTKIQVKME KLSSTTKGIC ELENYHYGEE SKRPPLFHTW PTTHFYEVSH KLLEMYRKEL LLKRTVAKEL AHTGDPDLTL SYLSMWLHQP YVESDSRLHL ESMLLETGHR AL
CINP Monoclonal antibody specifically detects CINP in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| CINP | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1810047K05Rik; 2810452K22Rik; CDK2-interacting protein; Cinp; cyclin dependent kinase 2 interacting protein; cyclin-dependent kinase 2 interacting protein; cyclin-dependent kinase 2-interacting protein; MGC849; RGD1563395 | |
| Recombinant human CINP (immunogen range is 1-212). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CINP |
| K70038_12G9 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BW66 | |
| 51550 | |
| IgG2a | |
| Protein A | |
| Antibody |