missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CINP Monoclonal Antibody (K70038_12G9), Invitrogen™

Catalog No. MA556111
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556111 200 μL
MA555731 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556111 Supplier Invitrogen™ Supplier No. MA556111
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MEAKTLGTVT PRKPVLSVSA RKIKDNAADW HNLILKWETL NDAGFTTANN IANLKISLLN KDKIELDSSS PASKENEEKV CLEYNEELEK LCEELQATLD GLTKIQVKME KLSSTTKGIC ELENYHYGEE SKRPPLFHTW PTTHFYEVSH KLLEMYRKEL LLKRTVAKEL AHTGDPDLTL SYLSMWLHQP YVESDSRLHL ESMLLETGHR AL

The function of this gene has not been reported. This gene contains a trinucleotide (ctt) repeat length polymorphism; alleles with either one or two repeat units have been identified.

Specifications

Antigen CINP
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K70038_12G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CINP
Gene Accession No. Q9BW66
Gene Alias 1810047K05Rik; 2810452K22Rik; CDK2-interacting protein; Cinp; cyclin dependent kinase 2 interacting protein; cyclin-dependent kinase 2 interacting protein; cyclin-dependent kinase 2-interacting protein; MGC849; RGD1563395
Gene Symbols CINP
Host Species Mouse
Immunogen Recombinant human CINP (immunogen range is 1-212).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less