missing translation for 'onlineSavingsMsg'
Learn More

Cinnamoyl chloride, 98%, predominantly trans

Catalog No. AC110390050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110390050 5 g Glass bottle
AC110391000 100 g Glass bottle
AC110395000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110390050 Supplier Thermo Scientific Chemicals Supplier No. 110390050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 102-92-1 | C9H7ClO | 166.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-92-1
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.61
MDL Number MFCD00000732
InChI Key WOGITNXCNOTRLK-VOTSOKGWSA-N
Synonym cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
PubChem CID 5354261
IUPAC Name (E)-3-phenylprop-2-enoyl chloride
SMILES C1=CC=C(C=C1)C=CC(=O)Cl

Specifications

Melting Point 33.0°C to 37.0°C
Color White to Yellow
Boiling Point 256.0°C to 258.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH=CHCOCl
Quantity 5 g
Merck Index 15, 2301
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in hydrocarbons and esters,soluble in dioxane,very soluble in logroin
Formula Weight 166.61
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material Cinnamoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 203-065-5
TSCA TSCA

RUO – Research Use Only