CAS RN 102-92-1
IUPAC Name:
(2E)-3-phenylprop-2-enoyl chloride
Synonyms:
(2E)-3-phenylprop-2-enoyl chloride
Molecular Weight (g/mol):
166.6
Molecular Formula:
C9H7ClO
InChi Key:
WOGITNXCNOTRLK-VOTSOKGWSA-N
SMILES:
ClC(=O)\C=C\C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Cinnamoyl chloride, 98%, predominantly trans
Cinnamoyl chloride, 98%, predominantly trans
CAS: 102-92-1 Molecular Formula: C9H7ClO Molecular Weight (g/mol): 166.61 MDL Number: MFCD00000732 InChI Key: WOGITNXCNOTRLK-VOTSOKGWSA-N Synonym: cinnamoyl chloride,cinnamic acid chloride,cinnamic chloride,trans-3-phenylacryloyl chloride,2-propenoyl chloride, 3-phenyl,cinnamoylchloride,e-cinnamoyl chloride,trans-cinnamoyl chloride,2e-3-phenylprop-2-enoyl chloride,3-phenyl-2-propenoyl chloride PubChem CID: 5354261 IUPAC Name: (E)-3-phenylprop-2-enoyl chloride SMILES: C1=CC=C(C=C1)C=CC(=O)Cl