missing translation for 'onlineSavingsMsg'
Learn More

Cinnamonitrile, 97%, predominantly trans

Catalog No. AC110380050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110380050 5 g Glass bottle
AC110380250 25 g Glass bottle
AC110381000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110380050 Supplier Thermo Scientific Chemicals Supplier No. 110380050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1885-38-7 | C9H7N | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1885-38-7
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00001930
InChI Key ZWKNLRXFUTWSOY-QPJJXVBHSA-N
Synonym cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
PubChem CID 1550846
IUPAC Name (E)-3-phenylprop-2-enenitrile
SMILES C1=CC=C(C=C1)C=CC#N

Specifications

Melting Point 20°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 254°C to 255°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 10% max. (cis-Cinnamonitrile) (GC)
Packaging Glass bottle
Refractive Index 1.6000 to 1.602
Linear Formula C6H5CH=CHCN
Quantity 5 g
Beilstein 09, 589
Specific Gravity 1.02
Solubility Information Solubility in water: insoluble
Formula Weight 129.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cinnamonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 217-552-5
TSCA TSCA

RUO – Research Use Only