missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinnamonitrile, 97%, predominantly trans
Description
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air & light sensitive. Store in dark. Incompatible with oxidizing agents.
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air & light sensitive. Store in dark. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 18°C to 20°C |
| Density | 1.029 |
| Boiling Point | 253°C to 254°C |
| Flash Point | 126°C (259°F) |
| Refractive Index | 1.602 |
| Quantity | 25 g |
| Sensitivity | Air and light sensitive |
| Solubility Information | Soluble in alcohol. Insoluble in water. |
| Formula Weight | 129.16 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H302-H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 217-552-5 |
| RTECSNumber | UD1440000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only