missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinnamamide, 97%, predominantly trans, Thermo Scientific™
$106.23 - $434.20
Chemical Identifiers
| CAS | 621-79-4 |
|---|---|
| Molecular Formula | C9H9NO |
| Molecular Weight (g/mol) | 147.18 |
| MDL Number | MFCD00008033 |
| InChI Key | APEJMQOBVMLION-VOTSOKGWSA-N |
| Synonym | cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine |
| PubChem CID | 5273472 |
| ChEBI | CHEBI:76320 |
| IUPAC Name | (E)-3-phenylprop-2-enamide |
| SMILES | C1=CC=C(C=C1)C=CC(=O)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC110360050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC110360250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C9H9NO | |
| MFCD00008033 | |
| cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine | |
| CHEBI:76320 | |
| C1=CC=C(C=C1)C=CC(=O)N |
Specifications
| 621-79-4 | |
| 96% min. (HPLC) | |
| C6H5CH=CHCONH2 | |
| 09, 587 | |
| APEJMQOBVMLION-VOTSOKGWSA-N | |
| (E)-3-phenylprop-2-enamide | |
| 5273472 | |
| 147.18 | |
| Cinnamamide, Predominantly trans |
| Authentic | |
| C9H9NO | |
| MFCD00008033 | |
| cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine | |
| C1=CC=C(C=C1)C=CC(=O)N | |
| 147.18 | |
| CHEBI:76320 | |
| 97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 210-707-8
RUO – Research Use Only