missing translation for 'onlineSavingsMsg'
Learn More

Cinnamamide, 97%, predominantly trans

Catalog No. AC110360050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110360050 5 g Glass bottle
AC110360250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110360050 Supplier Thermo Scientific Chemicals Supplier No. 110360050

CAS: 621-79-4 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-79-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00008033
InChI Key APEJMQOBVMLION-VOTSOKGWSA-N
Synonym cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine
PubChem CID 5273472
ChEBI CHEBI:76320
IUPAC Name (E)-3-phenylprop-2-enamide
SMILES C1=CC=C(C=C1)C=CC(=O)N

Specifications

Melting Point 146.0°C to 150.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCONH2
Quantity 5 g
Beilstein 09, 587
Formula Weight 147.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Cinnamamide, Predominantly trans
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 210-707-8

RUO – Research Use Only