missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cimetidine

Catalog No. p-7214745
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467520050 5 g
AC467520250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467520050 Supplier Thermo Scientific Chemicals Supplier No. 467520050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51481-61-9
Molecular Formula C10H16N6S

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: very slightly soluble aq. soln.. Other solubilities: soluble in methanol, glacial acetic acid, slightly soluble in ethanol, insoluble in ether
Melting Point 141.0°C to 143.0°C
Formula Weight 252.32
InChI Key AQIXAKUUQRKLND-UHFFFAOYSA-N
SMILES CN=C(NCCSCC1=C(C)NC=N1)NC#N
IUPAC Name N-cyano-N''-methyl-N'-(2-{[(5-methyl-1H-imidazol-4-yl)methyl]sulfanyl}ethyl)guanidine
Molecular Weight (g/mol) 252.34
Chemical Name or Material Cimetidine
EINECSNumber 257-232-2

RUO – Research Use Only