missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ CIM 0216
Description
CIM 0216 is a selective TRPM3 agonist. Exhibits selectivity for TRPM3 over TRPM1, TRPM2 and TRPM4-8. Stimulates insulin release from pancreatic islet cells in vitro. Evokes nocifensive behavior in mice.
Specifications
Specifications
| Chemical Name or Material | 3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-α-phenyl-1(2H)-quinolineacetamide |
| CAS | 1031496-06-6 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | TRPM Agonists |
| Molecular Formula | C21H21N3O2 |
| Formula Weight | 347.41 |
| Purity | >98% |