Learn More
Thermo Scientific™ Ciclopirox olamine
Chemical Identifiers
| CAS | 41621-49-2 |
|---|---|
| Molecular Formula | C14H24N2O3 |
| Molecular Weight (g/mol) | 268.36 |
| InChI Key | MBRHNTMUYWQHMR-UHFFFAOYSA-N |
| IUPAC Name | 2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one |
| SMILES | NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC463440010
|
Thermo Scientific™
463440010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC463440050
|
Thermo Scientific™
463440050 |
5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 41621-49-2 | |
| 268.36 | |
| 2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one |
| C14H24N2O3 | |
| MBRHNTMUYWQHMR-UHFFFAOYSA-N | |
| NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1 |
Specifications
| 41621-49-2 | |
| 1 g | |
| MBRHNTMUYWQHMR-UHFFFAOYSA-N | |
| 2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one | |
| 268.35 |
| C14H24N2O3 | |
| Solubility aq. soln.: .. Other solubilities: almost insoluble in dimethylsulfoxide and methanol | |
| NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1 | |
| 268.36 | |
| Ciclopirox olamine |
Safety and Handling
GHS H Statement Flammable solid.
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Very toxic to aquatic life
GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the risk of explosion.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 255-464-9
RUO â Research Use Only