accessibility menu, dialog, popup

UserName

CH3ONOHNH2HO

CAS RN 41621-49-2

IUPAC Name: 2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
Synonyms: ciclopirox olamine
Molecular Weight (g/mol): 268.36
Molecular Formula: C14H24N2O3
InChi Key: MBRHNTMUYWQHMR-UHFFFAOYSA-N
SMILES: NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)