missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ciclopirox olamine

Supplier:  Thermo Scientific™ 463440050

 View more versions of this product

Catalog No. AC463440050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ciclopirox olamine
C14H24N2O3
Solubility aq. soln.: .. Other solubilities: almost insoluble in dimethylsulfoxide and methanol
NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1
268.36
41621-49-2
5 g
MBRHNTMUYWQHMR-UHFFFAOYSA-N
2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
268.35

Chemical Identifiers

41621-49-2
268.36
2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
C14H24N2O3
MBRHNTMUYWQHMR-UHFFFAOYSA-N
NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Flammable solid.

May cause fire or explosion; strong oxidizer.

Causes severe skin burns and eye damage.

May cause an allergic skin reaction.

May cause respiratory irritation.

Very toxic to aquatic life

GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

Take any precaution to avoid mixing with combustibles.

In case of major fire and large quantities: Evacuate area.

Fight fire remotely due to the risk of explosion.

Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 255-464-9

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only