missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CIA 1
Description
CIA 1 is an inhibitor of the nuclear receptor COUP-TFII (IC50 ranged from 1.2 μM to 7.6 μM, in prostate cancer cell lines). In vitro, CIA 1 inhibits growth of multiple prostate cancer cell lines and suppresses COUP-TFII activity to regulate its target genes. In vivo, CIA 1 reduces tumor growth in prostate cancer xenograft mouse models.
Specifications
Specifications
| Chemical Name or Material | N-[2-(3,4-Dimethoxyphenyl)ethyl]-6-methylthieno[2,3-d]pyrimidin-4-amine hydrochloride |
| CAS | 1049691-47-5 |
| Quantity | 10 mg |
| Target | Additional Nuclear Receptor Compounds |
| Molecular Formula | C17H19N3O2S.HCl |
| Purity | 0.98 |