missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chymotrypsin, Human Pancreas

Catalog No. 23090050UG
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23090050UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23090050UG Supplier MilliporeSigma™ Supplier No. 23090050UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV

  • Natiave chymotrypsin from human pancreas
  • Pancreatic digestive enzyme
  • Increased levels of this enzyme and/or its zymogen have been found in serum of individuals with cystic fibrosis

Specifications

Formulation Salt free
Purity ≥95% by SDS-PAGE
Content And Storage -20°C
Form Lyophilized
Quantity 50 μg
Solubility Ice Cold 1 mM HCl
Product Type Chymotrypsin, menschliche Pankreas