missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium ICP Standard, 1000 ppm Cr6+ in 3% HCl, Ricca Chemical

Catalog No. p-3340634
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PCR61KH100 Natural LDPE Bottle
PCR61KH500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PCR61KH100 Supplier Ricca Chemical Company Supplier No. PCR61KH100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7789-09-5 , 7647-01-0 , 7732-18-5
Molecular Formula Cr2H8N2O7
Molecular Weight (g/mol) 252.06
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
PubChem CID 24600
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 94.0, 3.34, 0.23
CAS Max % 98.0, 3.48, 0.24
Concentration or Composition (by Analyte or Components) 1mL = 1mg Cr (1, 000ppm Cr)(NH4)2Cr2O7 in 3% HCl
Chemical Name or Material Chromium(VI) ICP Standard
ShelfLife 18 months