missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexavalent Chromium ICP Standard, 10,000 ppm Cr6+ in 5% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 89.0, 6.7, 2.16 |
| CAS Max % | 93.0, 6.97, 2.25 |
| Concentration or Composition (by Analyte or Components) | 1mL = 10mg Cr (10, 000ppm Cr)(NH4)2Cr2O7 in 5% HNO3 |
| Chemical Name or Material | Chromium(VI) ICP Standard |
Safety and Handling
| ShelfLife | 18 months |