missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium ICP Standard, 10,000 ppm Cr6+ in 5% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PCR610KN100

Packaging Natural LDPE Bottle

 View more versions of this product

Catalog No. PCR610KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7789-09-5 , 7697-37-2 , 7732-18-5
252.06
24600
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
Cr2H8N2O7
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium

Specifications

Chromium(VI) ICP Standard
7789-09-5 , 7697-37-2 , 7732-18-5
93.0, 6.97, 2.25
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24600
Natural LDPE Bottle
89.0, 6.7, 2.16
Cr2H8N2O7
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
252.06
1mL = 10mg Cr (10, 000ppm Cr)(NH4)2Cr2O7 in 5% HNO3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents