missing translation for 'onlineSavingsMsg'
Learn More

Chromium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLCR22X

 View more versions of this product

Catalog No. PLCR2-2X


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cr1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Chromium Standard
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP
Certified Reference Material
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
500 mL
Assurance
Triple-leached LDPE bottle
7789-09-5 , 7732-18-5
Cr2H8N2O7
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
JOSWYUNQBRPBDN-UHFFFAOYSA-P
2% HNO3
252.06
24600

Chemical Identifiers

7789-09-5
252.06
24600
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
Cr2H8N2O7
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS Documents

Documents

Promotions

Promotions

Product intended for laboratory use only