missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride, 99%, anhydrous

CAS: 10025-73-7 | Cl3Cr | 158.36 g/mol

Supplier:  Thermo Scientific Chemicals 190470250

 View more versions of this product

Catalog No. AC190470250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chromium(III) chloride
99%
Purple
25 g
Cl3CrH12O6
MFCD00010948
15, 2226
Solubility in water: slightly soluble
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
266.44
99%
2.76
1152.0°C
10025-73-7
Shiny Fluffy Powder or Flakes
98.5% min
CrCl3
15,95
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium(3+) hexahydrate trichloride
158.36
Plastic bottle

Chemical Identifiers

10025-73-7
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium(3+) hexahydrate trichloride
MFCD00010948
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
May be corrosive to metals.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Absorb spillage to prevent material damage.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SK

GHS Signal Word: Warning

EINECSNumber : 233-038-3

RTECSNumber : GB5425000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only