missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride, 99%, anhydrous

Catalog No. p-222856
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190470050 5 g Glass bottle
AC190470250 25 g Plastic bottle
AC190471000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC190470050 Supplier Thermo Scientific Chemicals Supplier No. 190470050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10025-73-7 | Cl3Cr | 158.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-73-7
MDL Number MFCD00010948
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f
IUPAC Name chromium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

Melting Point 1152.0°C
Color Purple
Physical Form Shiny Fluffy Powder or Flakes
Quantity 5 g
Assay Percent Range 98.5% min
Molecular Formula Cl3CrH12O6
Linear Formula CrCl3
Fieser 15,95
Merck Index 15, 2226
Solubility Information Solubility in water: slightly soluble
Molecular Weight (g/mol) 266.44
Formula Weight 158.36
Percent Purity 99%
Packaging Glass bottle
Density 2.76
Chemical Name or Material Chromium(III) chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
May be corrosive to metals.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Absorb spillage to prevent material damage.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SK
EINECSNumber 233-038-3
RTECSNumber GB5425000
TSCA TSCA

RUO – Research Use Only