missing translation for 'onlineSavingsMsg'
Learn More

Chromium AA Standard, 1000 ppm Cr in 3% HCl, Ricca Chemical

Catalog No. ACR1KH500
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ACR1KH500 500 mL Natural Poly Bottle
ACR1KH100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ACR1KH500 Supplier Ricca Chemical Company Supplier No. ACR1KH500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7789-09-5 | Formula: Cr2H8N2O7 | Weight: 252.06 g/mol

Chemical Identifiers

CAS 7789-09-5 , 7732-18-5 , 7789-09-5
Molecular Formula Cr2H8N2O7
Molecular Weight (g/mol) 252.06
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
PubChem CID 24600
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm Cr
CAS Min % 97.0, 1.12, 0.22
CAS Max % 100.0, 1.16, 0.23
Equivalence 1 mL = 1 mg Cr
Physical Form Liquid
Color Blue
Boiling Point 100°C
Type Single-element Standard Solution
Quantity 500 mL
Grade High Purity
Chemical Name or Material Chromium AA Standard
Show More Show Less