missing translation for 'onlineSavingsMsg'
Learn More

Ahlstrom-Munksjo Chromatography, Electrophoresis and Blotting Paper, Grade 238

Medium thick paper is recommended for general chromatography application. Also ideal for use as wicks in protein or nucleic acid blotting to provide a uniform flow or buffer through the gel to transfer membrane in a blotting sandwich.

$82.97 - $908.21

Specifications

For Use With (Application) Chromatography applications. Use as wicks in protein or nucleic acid blotting. Suited for electrophoresis
Grade 238
Thickness (Metric) 0.35 mm
View More Specs

Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
09-790-53B
View Documents
Ahlstrom-Munksjo
23880710
70 x 100 mm
Pack of 100 Each for $82.97
Only null left
 
09-790-53G
View Documents
Ahlstrom-Munksjo
23882020
200 x 200 mm
Pack of 100 Each for $253.88
Only null left
 
22-279539
View Documents
Ahlstrom-Munksjo
23884657
460 x 570 mm
Pack of 100 Each for $908.21
Only null left
 
Description

Description

Specifications

Specifications

Chromatography applications. Use as wicks in protein or nucleic acid blotting. Suited for electrophoresis
238
0.35 mm
Videos
SDS
Documents

Documents