Learn More
Ahlstrom-Munksjo Chromatography, Electrophoresis and Blotting Paper, Grade 238
Medium thick paper is recommended for general chromatography application. Also ideal for use as wicks in protein or nucleic acid blotting to provide a uniform flow or buffer through the gel to transfer membrane in a blotting sandwich.
Supplier:  Ahlstrom-Munksjo 23880710
 View more versions of this product
Catalog No. 09-790-53B
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Chromatography applications. Use as wicks in protein or nucleic acid blotting. Suited for electrophoresis
0.35 mm
70 mm
238
70 x 100 mm
100 mm
Product Suggestions
Product Suggestions