missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromate Standard, 1000 ppm CrO42-, Ricca Chemical
Supplier: Ricca Chemical Company 207016
| Packaging | Natural Poly Bottle |
|---|
Chemical Identifiers
| 7789-00-6 , 7732-18-5 | |
| 194.19 | |
| XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
| CHEBI:75249 | |
| [K+].[K+].[O-][Cr]([O-])(=O)=O |
| CrK2O4 | |
| MFCD00011368 | |
| 24597 | |
| dipotassium dioxochromiumbis(olate) |
Specifications
| Chromate Standard | |
| 7789-00-6 , 7732-18-5 | |
| 100.0, 0.17 | |
| CrK2O4 | |
| XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
| dipotassium dioxochromiumbis(olate) | |
| 24597 |
| Natural Poly Bottle | |
| 98.0, 0.17 | |
| 1 mL = 1 mg CrO | |
| MFCD00011368 | |
| [K+].[K+].[O-][Cr]([O-])(=O)=O | |
| 194.19 | |
| CHEBI:75249 |