missing translation for 'onlineSavingsMsg'
Learn More

Choline Dihydrogen Citrate, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C122925KG

 View more versions of this product

Catalog No. 18609728


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

77-91-8
Poly Pail
2.5 kg
C[N+](C)(C)CCO.OC(=O)CC(O)(CC(O)=O)C([O-])=O
295.29
Reagent
1
C11H21NO8
WRPUOFKIGGWQIJ-UHFFFAOYSA-M
(2-hydroxyethyl)trimethylazanium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
0.98

Chemical Identifiers

77-91-8
295.29
(2-hydroxyethyl)trimethylazanium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
C11H21NO8
WRPUOFKIGGWQIJ-UHFFFAOYSA-M
C[N+](C)(C)CCO.OC(=O)CC(O)(CC(O)=O)C([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.