missing translation for 'onlineSavingsMsg'
Learn More

Choline Dihydrogen Citrate, 98%, Spectrum™ Chemical

Catalog No. p-7165317
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609731 500 g Poly Pail
18609728 2.5 kg Poly Pail
18609729 12 kg Poly Pail
18609730 25 kg Fiber Drum
18609732 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609731 Supplier Spectrum Chemical Mfg Cor Supplier No. C1229500GM
Only null left

Chemical Identifiers

CAS 77-91-8
Molecular Formula C11H21NO8
Molecular Weight (g/mol) 295.29
InChI Key WRPUOFKIGGWQIJ-UHFFFAOYSA-M
IUPAC Name (2-hydroxyethyl)trimethylazanium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
SMILES C[N+](C)(C)CCO.OC(=O)CC(O)(CC(O)=O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 500 g
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.