accessibility menu, dialog, popup

UserName

CAS RN 77-91-8

CH3NCH3CH3HOHOOOHOHOOO

CAS RN 77-91-8

IUPAC Name: (2-hydroxyethyl)trimethylazanium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
Synonyms: choline 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
Molecular Weight (g/mol): 295.29
Molecular Formula: C11H21NO8
InChi Key: WRPUOFKIGGWQIJ-UHFFFAOYSA-M
SMILES: C[N+](C)(C)CCO.OC(=O)CC(O)(CC(O)=O)C([O-])=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)