missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholesterol, NF, 95-102%, Spectrum™ Chemical
CH135, 57-88-5, C27H46O
$309.89 - $62958.86
Chemical Identifiers
| CAS | 57-88-5 |
|---|---|
| Molecular Formula | C27H46O |
| Molecular Weight (g/mol) | 386.66 |
| InChI Key | HVYWMOMLDIMFJA-DPAQBDIFSA-N |
| IUPAC Name | (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol |
| SMILES | CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601652
|
Spectrum Chemical Mfg Cor
CH13525GM |
25 g | Amber Glass Bottle |
Each for $309.89
|
|
||||
|
18601653
|
Spectrum Chemical Mfg Cor
CH135125GM |
125 g | Amber Glass Bottle |
Each for $772.29
|
|
||||
|
18601654
|
Spectrum Chemical Mfg Cor
CH135500GM |
500 g | Amber Glass Bottle |
Each for $2,218.03
|
|
||||
|
18601655
|
Spectrum Chemical Mfg Cor
CH13525KG |
2.5 kg | Poly Pail |
Each for $6,970.74
|
|
||||
|
18601656
|
Spectrum Chemical Mfg Cor
CH13550KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Cholesterol, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 57-88-5 | |
| 386.66 | |
| (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol |
| C27H46O | |
| HVYWMOMLDIMFJA-DPAQBDIFSA-N | |
| CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
Specifications
| 57-88-5 | |
| 0.1% | |
| 147-150°C | |
| C27H46O | |
| -34° to -38° | |
| CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C | |
| 386.66 | |
| NF |
| 100% | |
| 0.3% | |
| Amber Glass Bottle | |
| 25 g | |
| HVYWMOMLDIMFJA-DPAQBDIFSA-N | |
| (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol | |
| 95-102% |